4-Chloro-7-(trifluoromethyl)quinoline 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-7-trifluoromethylquinoline

A175245-10G A175245-1G A175245-250MG A175245-25G A175245-5G
76799-514EA 90.18 USD
76799-514 76799-516 76799-518 76799-520 76799-522
4-Chloro-7-(trifluoromethyl)quinoline 98%
4-Chloro-7-(trifluoromethyl)quinoline

Precaución: for Research Use Only

Formula: C₁₀H₅ClF₃N
MW: 231.6016 g/mol
MDL Number: MFCD00006775
CAS Number: 346-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR