DL-O-Methylserine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-3-methoxypropanoic acid

A174606-1G A174606-250MG A174606-25G A174606-5G
76799-488EA 46.68 USD
76799-488 76799-490 76799-492 76799-494
DL-O-Methylserine 97%
DL-O-Methylserine

Precaución: for Research Use Only

Formula: C₄H₉NO₃
MW: 119.12 g/mol
MDL Number: MFCD00037773
CAS Number: 19794-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR