Histamine diphosphate 97%

Supplier: Ambeed
Danger

A174108-250MG A174108-1MG A174108-5G A174108-25G
76799-482EA 9 USD
76799-482 77667-426 77667-428 77879-148
Histamine diphosphate 97%
Histamine diphosphate

Precaución: for Research Use Only

Formula: C₅H₉N₃·2H₃PO₄
MDL Number: MFCD00064316
CAS Number: 51-74-1
UN: 3259
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR