2-Chloroquinoline-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A173688-10G A173688-1G A173688-250MG A173688-5G A173688-25G
76799-470EA 45 USD
76799-470 76799-472 76799-474 76799-476 77550-120
2-Chloroquinoline-4-carboxylic acid 97%
2-Chloroquinoline-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆ClNO₂
MDL Number: MFCD00023940
CAS Number: 5467-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR