Sodium 1-Naphthalenesulfonate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Naphthalenesulphonic acid sodium salt, Naphthalene-1-sulphonic acid sodium salt
A173402-100G
A173402-25G
A173402-500G
76799-458EA
39.26
USD
76799-458
76799-460
76799-462
Sodium 1-Naphthalenesulfonate 98%
Sodium 1-Naphthalenesulfonate
Precaución: for Research Use Only
Formula:
C₁₀H₈O₃S MW: 230.21 g/mol |
MDL Number:
MFCD00064964 CAS Number: 130-14-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...