Ethyl-2-methyl-4-pentenoate ≥98% stabilized

Supplier: Ambeed
Danger

Synonyms: Ethyl 2-methylpent-4-enoate, 2-Methyl-4-pentenoic Acid Ethyl Ester

Stabilization: Stabilized with Tocopherols 0.2%

A172917-5G A172917-25G
76799-456EA 10.61 USD
76799-456 77567-546
Ethyl-2-methyl-4-pentenoate ≥98% stabilized
Ethyl-2-methyl-4-pentenoate

Precaución: For research use only.

Formula: C₈H₁₄O₂
MW: 142.20 g/mol
MDL Number: MFCD00010235
CAS Number: 53399-81-8
UN: 3272
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR