Ethyl-4-cyano-2-pyridinecarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-cyanopicolinate

A172855-100MG A172855-250MG
76799-452EA 87 USD
76799-452 76799-454
Ethyl-4-cyano-2-pyridinecarboxylate 97%
Ethyl-4-cyano-2-pyridinecarboxylate

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MW: 176.18 g/mol
MDL Number: MFCD09152770
CAS Number: 97316-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR