Lithium acetate 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
Acetic acid lithium salt, Lithiumacetate
A172324-100G
A172324-25G
A172324-500G
76799-446EA
36
USD
76799-446
76799-448
76799-450
Lithium acetate 99%
Lithium acetate
Precaución: for Research Use Only
Formula:
H₃CCOOLi MW: 65.99 g/mol Melting Pt: 291 |
MDL Number:
MFCD00013057 CAS Number: 546-89-4 Merck Index: 12,05544 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...