Iron(III) acetylacetonate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Tris(2,4-pentanedionato-O,O')iron (III), Ferric acetylacetonate, Iron (III) 2,4-pentanedionate, Iron (III) tris(2,4-pentanedionato-O,O')
A171511-1000G
A171511-100G
A171511-25G
A171511-500G
76799-432EA
55
USD
76799-432
76799-434
76799-436
76799-438
Iron(III) acetylacetonate 97%
Iron(III) acetylacetonate
Precaución: for Research Use Only
Formula:
C₁₅H₂₁FeO₆ MW: 353.18 g/mol Boiling Pt: 100 |
MDL Number:
MFCD00000020 CAS Number: 14024-18-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...