KN-93 98%

Supplier: Ambeed
Warning

A170870-10MG A170870-1MG A170870-25MG A170870-50MG A170870-5MG
76799-390EA 146.41 USD
76799-390 76799-392 76799-394 76799-396 76799-398
KN-93 98%
KN-93

Precaución: for Research Use Only

Formula: C₂₆H₂₉ClN₂O₄S
MW: 501.04 g/mol
MDL Number: MFCD00236424
CAS Number: 139298-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR