N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-alanine pentafluorophenyl ester 98% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: Fmoc-Ala-OPfp, N-Fmoc-L-alanine Pentafluorophenyl Ester, Fmoc-Ala-Opfp

A170738-1G A170738-250MG A170738-25G A170738-5G
76799-380EA 31.83 USD
76799-380 76799-382 76799-384 76799-386
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-alanine pentafluorophenyl ester 98% (by HPLC)
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-alanine pentafluorophenyl ester

Precaución: for Research Use Only

Formula: C₂₄H₁₆F₅NO₄
MW: 477.39 g/mol
MDL Number: MFCD00065612
CAS Number: 86060-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR