7-Methoxybenzofuran-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A169513-1G A169513-25G A169513-5G
76799-344EA 15.92 USD
76799-344 76799-346 76799-348
7-Methoxybenzofuran-2-carboxylic acid 98%
7-Methoxybenzofuran-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₈O₄
MW: 192.17 g/mol
MDL Number: MFCD00060516
CAS Number: 4790-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR