Ethyl-3-bromo-2-(bromomethyl)propionate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-bromo-2-(bromomethyl)propanoate, 3-Bromo-2-(bromomethyl)propionic Acid Ethyl Ester

A167943-1G A167943-250MG A167943-25G A167943-5G
76799-280EA 22.28 USD
76799-280 76799-282 76799-284 76799-286
Ethyl-3-bromo-2-(bromomethyl)propionate 98%
Ethyl-3-bromo-2-(bromomethyl)propionate

Precaución: for Research Use Only

Formula: C₆H₁₀Br₂O₂
MW: 273.95 g/mol
MDL Number: MFCD00034154
CAS Number: 58539-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR