3-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-1-propanol 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-β-Ala-ol, 3-(Fmoc-amino)-1-propanol

A167565-100G A167565-1G A167565-250MG A167565-5G
76799-272EA 506.05 USD
76799-272 76799-274 76799-276 76799-278
3-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-1-propanol 97%
3-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-1-propanol

Precaución: for Research Use Only

Formula: C₁₈H₁₉NO₃
MW: 297.35 g/mol
MDL Number: MFCD00409484
CAS Number: 157887-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR