Dimethyl glutarate 98%

Supplier: Ambeed
Warning

Synonyms: 1,5-Pentanedioic acid dimethyl ester, Glutaric acid dimethyl ester

A166537-1000G A166537-100G A166537-25G A166537-500G
76799-224EA 66.84 USD
76799-224 76799-226 76799-228 76799-230
Dimethyl glutarate 98%
Dimethyl glutarate

Precaución: for Research Use Only

Formula: C₇H₁₂O₄
MW: 160.17 g/mol
MDL Number: MFCD00008468
CAS Number: 1119-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR