tert-Butyl-3-(hydroxymethyl)piperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-(hydroxymethyl)piperidine-1-carboxylate, 3-(Hydroxymethyl)piperidine, N-BOC protected

A166519-100G A166519-10G A166519-25G A166519-5G
76799-216EA 75 USD
76799-216 76799-218 76799-220 76799-222
tert-Butyl-3-(hydroxymethyl)piperidine-1-carboxylate 95%
tert-Butyl-3-(hydroxymethyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₃
MDL Number: MFCD03094733
CAS Number: 116574-71-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR