Ethyl phenylpropiolate 95%

Supplier: Ambeed
Warning

A166265-1G A166265-25G A166265-5G
76799-206EA 7 USD
76799-206 76799-208 76799-210
Ethyl phenylpropiolate 95%
Ethyl phenylpropiolate

Precaución: for Research Use Only

Formula: C₁₁H₁₀O₂
MDL Number: MFCD00009185
CAS Number: 2216-94-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR