H-d-Ser(tBu)-OMe·HCl 97%

Supplier: Ambeed
Warning

Synonyms: O-tert-Butyl-D-serine methyl ester hydrochloride, H-D-Ser(tBu)-OMe·HCl, H-D-Ser(tBu)-OMe.HCl

A166026-10G A166026-250MG A166026-25G A166026-5G
76799-196EA 70 USD
76799-196 76799-198 76799-200 76799-202
H-d-Ser(tBu)-OMe·HCl 97%
H-d-Ser(tBu)-OMe·HCl

Precaución: for Research Use Only

Formula: C₈H₁₈ClNO₃
MW: 211.686 g/mol
MDL Number: MFCD00080901
CAS Number: 78537-14-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR