Fmoc-O-methyl-D-tyrosine 97%

Supplier: Ambeed
Warning

Synonyms: N-Fmoc-O-methyl-D-tyrosine, Fmoc-D-Tyr(Me)-OH

A165762-1G A165762-25G A165762-5G
76799-186EA 21 USD
76799-186 76799-188 76799-190 77567-412
Fmoc-O-methyl-D-tyrosine 97%
Fmoc-O-methyl-D-tyrosine

Precaución: For research use only.

Formula: C₂₅H₂₃NO₅
MW: 417.461 g/mol
MDL Number: MFCD00237393
CAS Number: 201335-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR