Dicyclopentyl(dimethoxy)silane 98%

Supplier: Ambeed
Warning

Synonyms: Dicyclopentyldimethoxysilane

A163924-100G A163924-10G A163924-1G A163924-25G A163924-5G
76799-114EA 480.59 USD
76799-114 76799-116 76799-118 76799-120 76799-122
Dicyclopentyl(dimethoxy)silane 98%
Dicyclopentyl(dimethoxy)silane

Precaución: for Research Use Only

Formula: C₁₂H₂₄O₂Si
MW: 228.41 g/mol
MDL Number: MFCD01074471
CAS Number: 126990-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR