ITE ≥98%

Supplier: Ambeed
Warning

Synonyms: Aryl hydrocarbon receptor ligand, 2-(1H-Indol-3-ylcarbonyl)-4-thiazolecarboxylic acid methyl ester, Methyl 2-(1H-indole-3-carbonyl)thiazole-4-carboxylate

A163287-10MG A163287-1MG A163287-5MG A163287-100MG A163287-250MG
76799-096EA 35.01 USD
76799-096 76799-098 76799-100 77667-282 77667-284
ITE ≥98%
ITE

Precaución: for Research Use Only

Formula: C₁₄H₁₀N₂O₃S
MW: 286.3 g/mol
MDL Number: MFCD06411597
CAS Number: 448906-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR