Glycyl-D-tryptophan 97%

Supplier: Ambeed
Warning

A163220-100MG A163220-1G A163220-250MG
76799-084EA 50.93 USD
76799-084 76799-086 76799-088
Glycyl-D-tryptophan 97%
Glycyl-D-tryptophan

Precaución: for Research Use Only

Formula: C₁₃H₁₅N₃O₃
MW: 261.28 g/mol
MDL Number: MFCD00065278
CAS Number: 50632-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR