N,N-Diethylbenzamide 98%

Supplier: Ambeed
Warning

A162722-100G A162722-25G A162722-5G
76799-078EA 155 USD
76799-078 76799-080 76799-082
N,N-Diethylbenzamide 98%
N,N-Diethylbenzamide

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO
MW: 177.25 g/mol
MDL Number: MFCD00026726
CAS Number: 1696-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR