Dibutyl carbonate 97%

Supplier: Ambeed
Warning

A162596-100G A162596-1G A162596-500G A162596-5G
76799-070EA 17 USD
76799-070 76799-072 76799-074 76799-076
Dibutyl carbonate 97%
Dibutyl carbonate

Precaución: for Research Use Only

Formula: C₉H₁₈O₃
MDL Number: MFCD00043938
CAS Number: 542-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR