D-(-)-Luciferin potassium salt 97%

Supplier: Ambeed
Warning

A162108-100MG A162108-10MG A162108-1G A162108-1MG A162108-250MG A162108-25MG A162108-5G
76799-036EA 31 USD
76799-036 76799-038 76799-040 76799-042 76799-044 76799-046 77567-368
D-(-)-Luciferin potassium salt 97%
D(-)-Luciferin potassium salt

Precaución: For research use only.

Formula: C₁₁H₇KN₂O₃S₂
MDL Number: MFCD00044928
CAS Number: 115144-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR