Ethyl-5-hydroxyindole-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-hydroxyindole-2-carboxylate

A161982-100MG A161982-250MG A161982-5G A161982-1G
76799-030EA 15.92 USD
76799-030 76799-032 76799-034 77600-298
Ethyl-5-hydroxyindole-2-carboxylate 97%
Ethyl-5-hydroxyindole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₃
MW: 205.213 g/mol
MDL Number: MFCD00015458
CAS Number: 24985-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR