5-Bromo-1-tetralone 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-3,4-dihydronaphthalen-1(2H)-one

A160498-10G A160498-1G A160498-5G A160498-25G A160498-250MG
76798-998EA 209 USD
76798-998 76799-000 76799-002 77592-956 77667-252
5-Bromo-1-tetralone 97%
5-Bromo-1-tetralone

Precaución: for Research Use Only

Formula: C₁₀H₉BrO
MW: 225.08 g/mol
MDL Number: MFCD02179288
CAS Number: 68449-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR