H-Thr(Ac)-OH 95%

Supplier: Ambeed
Warning

Synonyms: (2S,3R)-3-Acetoxy-2-aminobutanoic acid

A159972-1G A159972-250MG
76798-980EA 87 USD
76798-980 76798-982
H-Thr(Ac)-OH 95%
H-Thr(Ac)-OH

Precaución: for Research Use Only

Formula: C₆H₁₁NO₄
MW: 161.2 g/mol
MDL Number: MFCD00190723
CAS Number: 17012-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR