2-Hydroxy-4-(trifluoromethyl)quinoline 95%

Supplier: Ambeed
Warning

Synonyms: 4-Trifluoromethyl-2(1H)-quinolinone

A159518-1G A159518-250MG A159518-25G A159518-5G
76798-958EA 85.94 USD
76798-958 76798-960 76798-962 76798-964
2-Hydroxy-4-(trifluoromethyl)quinoline 95%
2-Hydroxy-4-(trifluoromethyl)quinoline

Precaución: for Research Use Only

Formula: C₁₀H₆F₃NO
MW: 213.16 g/mol
MDL Number: MFCD03407380
CAS Number: 25199-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR