Ethyl-6-chloroimidazo[1,2-b]pyridazine-2-carboxylate 98%

Supplier: Ambeed
Warning

A159470-100MG A159470-1G A159470-250MG A159470-5G A159470-25G
76798-950EA 6 USD
76798-950 76798-952 76798-954 76798-956 77879-092
Ethyl-6-chloroimidazo[1,2-b]pyridazine-2-carboxylate 98%
Ethyl 6-chloroimidazo[1,2-b]pyridazine-2-carboxylate

Precaución: for Research Use Only

Formula: C₉H₈ClN₃O₂
MDL Number: MFCD03086230
CAS Number: 64067-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR