3-((4-Nitrobenzyl)oxy)-3-oxopropanoic acid 97%

Supplier: Ambeed
Warning

A159452-100G A159452-10G A159452-25G A159452-500G A159452-5G
76798-940EA 68.96 USD
76798-940 76798-942 76798-944 76798-946 76798-948
3-((4-Nitrobenzyl)oxy)-3-oxopropanoic acid 97%
3-((4-Nitrobenzyl)oxy)-3-oxopropanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₉NO₆
MDL Number: MFCD00191556
CAS Number: 77359-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR