Isoindolin-4-amine dihydrochloride 97%

Supplier: Ambeed
Warning

A159156-100MG A159156-1G A159156-250MG
76798-928EA 147.47 USD
76798-928 76798-930 76798-932
Isoindolin-4-amine dihydrochloride 97%
Isoindolin-4-amine dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₂Cl₂N₂
MW: 207.10 g/mol
MDL Number: MFCD11053657
CAS Number: 92259-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR