2-Methoxyphenyl 2-acetoxybenzoate 95%

Supplier: Ambeed
Warning

Synonyms: Guacetisal

A158767-100MG A158767-10MG A158767-1MG A158767-25MG A158767-50MG A158767-5MG
76798-898EA 38.2 USD
76798-898 76798-900 76798-902 76798-904 76798-906 76798-908
2-Methoxyphenyl 2-acetoxybenzoate 95%
2-Methoxyphenyl 2-acetoxybenzoate

Precaución: for Research Use Only

Formula: C₁₆H₁₄O₅
MW: 286.27 g/mol
MDL Number: MFCD00864354
CAS Number: 55482-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR