Methyl-DL-2-aminobutyrate hydrochloride 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-Abu-OMe·HCl, DL-2-Aminobutyric Acid Methyl Ester Hydrochloride, Methyl 2-aminobutanoate hydrochloride
A158598-100G
A158598-10G
A158598-1G
A158598-25G
A158598-5G
A158598-500G
76798-888EA
53
USD
76798-888
76798-890
76798-892
76798-894
76798-896
77879-088
Methyl-DL-2-aminobutyrate hydrochloride 95%
Methyl-DL-2-aminobutyrate hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₁₁NO₂ |
MDL Number:
MFCD00058295 CAS Number: 7682-18-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...