3-(4-Methoxybenzoyl)propionic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-(4-Methoxyphenyl)-4-oxobutanoic acid

A157953-10G A157953-1G A157953-25G A157953-5G
76798-866EA 15 USD
76798-866 76798-868 76798-870 76798-872
3-(4-Methoxybenzoyl)propionic acid 98%
3-(4-Methoxybenzoyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₄
MW: 208.21 g/mol
MDL Number: MFCD00002795
CAS Number: 3153-44-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR