Diethyl-4-chloro-2,6-pyridinedicarboxylate 98%

Supplier: Ambeed
Warning

A157499-1G A157499-25G A157499-5G
76798-846EA 12 USD
76798-846 76798-848 76798-850
Diethyl-4-chloro-2,6-pyridinedicarboxylate 98%
Diethyl-4-chloro-2,6-pyridinedicarboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₂ClNO₄
MDL Number: MFCD00215521
CAS Number: 53389-01-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR