Maltohexaose 95%

Supplier: Ambeed
Warning

Synonyms: (2R,3R,4R,5R)-4-(((2R,3R,4R,5S,6R)-5-(((2R,3R,4R,5S,6R)-5-(((2R,3R,4R,5S,6R)-5-(((2R,3R,4R,5S,6R)-3,4-dihydroxy-6-(hydroxymethyl)-5-(((2R,3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2,3,5,6-tetrahydroxyhexanal

A156522-100MG A156522-250MG A156522-50MG A156522-1G
76798-822EA 106 USD
76798-822 76798-824 77667-214 77667-212
Maltohexaose 95%
Maltohexaose

Precaución: for Research Use Only

Formula: C₃₆H₆₂O₃₁
MDL Number: MFCD00083650
CAS Number: 34620-77-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR