DL-4-Hydroxymandelic acid monohydrate 95+%

Supplier: Ambeed
Warning

Synonyms: DL-alpha,4-Dihydroxyphenylacetic Acid, 2-Hydroxy-2-(4-hydroxyphenyl)acetic acid hydrate

A156212-25G A156212-5G A156212-100G A156212-500G
76798-814EA 81 USD
76798-814 76798-816 77879-078 77879-079
DL-4-Hydroxymandelic acid monohydrate 95+%
DL-4-Hydroxymandelic acid monohydrate

Precaución: for Research Use Only

Formula: C₈H₁₀O₅
MW: 186.16 g/mol
MDL Number: MFCD00149283
CAS Number: 184901-84-6

Order Now

Specification Test Results

Loss on drying ≤ 20%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR