H-D-Glu(OBzl)-OtBu·HCl 97%

Supplier: Ambeed
Warning

Synonyms: (R)-5-Benzyl 1-tert-butyl 2-aminopentanedioate hydrochloride

A156133-1G A156133-250MG
76798-810EA 24 USD
76798-810 76798-812
H-D-Glu(OBzl)-OtBu·HCl 97%
H-D-Glu(OBzl)-OtBu·HCl

Precaución: for Research Use Only

Formula: C₁₆H₂₃NO₄·HCl
MDL Number: MFCD04973241
CAS Number: 90159-60-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR