Ethyl-4-oxo-3-piperidinecarboxylate hydrochloride 97%

Supplier: Ambeed
Warning

A155946-1G A155946-5G A155946-25G A155946-100G
76798-790EA 5.31 USD
76798-790 76798-792 77600-228 77667-200
Ethyl-4-oxo-3-piperidinecarboxylate hydrochloride 97%
Ethyl-4-oxo-3-piperidinecarboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₃NO₃
MDL Number: MFCD00043267
CAS Number: 4644-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR