(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(pyridin-3-yl)propanoic acid 95%

Supplier: Ambeed
Warning

A154684-100MG A154684-250MG
76798-762EA 65 USD
76798-762 76798-764
(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(pyridin-3-yl)propanoic acid 95%
(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(pyridin-3-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₂₃H₂₀N₂O₄
MW: 388.42 g/mol
MDL Number: MFCD03428024
CAS Number: 511272-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR