Ethyl 2,3-Dicyanopropionate 97%

Supplier: Ambeed
Warning

A153226-100G A153226-25G A153226-5G A153226-500G
76798-728EA 30 USD
76798-728 76798-730 76798-732 77879-058
Ethyl 2,3-Dicyanopropionate 97%
Ethyl 2,3-Dicyanopropionate

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
MDL Number: MFCD08061844
CAS Number: 40497-11-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR