Lornoxicam 98%

Supplier: Ambeed
Warning

A152949-100MG A152949-10MG A152949-1G A152949-250MG A152949-50MG
76798-718EA 9 USD
76798-718 76798-720 76798-722 76798-724 76798-726
Lornoxicam 98%
Lornoxicam

Precaución: for Research Use Only

Formula: C₁₃H₁₀ClN₃O₄S₂
MDL Number: MFCD00866163
CAS Number: 70374-39-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR