Ethyl 2-methyl-1H-indole-3-carboxylate 98%

Supplier: Ambeed
Warning

A152891-100MG A152891-10G A152891-1G A152891-250MG A152891-25G A152891-5G
76798-706EA 9.55 USD
76798-706 76798-708 76798-710 76798-712 76798-714 76798-716
Ethyl 2-methyl-1H-indole-3-carboxylate 98%
Ethyl 2-methyl-1H-indole-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO₂
MW: 203.23 g/mol
MDL Number: MFCD01863674
CAS Number: 53855-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR