Fmoc-3-(aminooxy)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Aoa, Fmoc-Aoa-OH

A152880-250MG A152880-1G A152880-5G
76798-704EA 46.68 USD
76798-704 77600-174 77667-162
Fmoc-3-(aminooxy)acetic acid 97%
Fmoc-3-(aminooxy)acetic acid

Precaución: for Research Use Only

Formula: C₁₇H₁₅NO₅
MW: 313.31 g/mol
MDL Number: MFCD01076133
CAS Number: 123106-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR