4-Chloro-6-(trifluoromethyl)quinoline 96%
Supplier: Ambeed
|
Warning
|
A151700-250MG
A151700-25G
A151700-5G
76798-668EA
18.04
USD
76798-668
76798-670
76798-672
4-Chloro-6-(trifluoromethyl)quinoline 96%
4-Chloro-6-(trifluoromethyl)quinoline
Precaución: for Research Use Only
Formula:
C₁₀H₅ClF₃N MW: 231.61 g/mol |
MDL Number:
MFCD00153106 CAS Number: 49713-56-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...