4-(Aminomethyl)benzenesulfonamide acetate 98%

Supplier: Ambeed
Warning

A151118-100G A151118-100MG A151118-10G A151118-1G A151118-25G A151118-5G
76798-654EA 144.29 USD
76798-654 76798-656 76798-658 76798-660 76798-662 76798-664
4-(Aminomethyl)benzenesulfonamide acetate 98%
4-(Aminomethyl)benzenesulfonamide acetate

Precaución: for Research Use Only

Formula: C₉H₁₄N₂O₄S
MW: 246.2835 g/mol
MDL Number: MFCD00072089
CAS Number: 13009-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR