(-)-Menthone 90% (by GC)

Supplier: Ambeed
Danger

A150418-100G A150418-25G A150418-500G A150418-1000G
76798-630EA 67 USD
76798-630 76798-632 76798-634 77879-042
(-)-Menthone 90% (by GC)
(-)-Menthone

Precaución: for Research Use Only

Formula: C₁₀H₁₈O
MDL Number: MFCD00001634
CAS Number: 14073-97-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR