Ethyl-3,6-dichloropyridazine-4-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3,6-dichloropyridazine-4-carboxylate

A150241-100MG A150241-1G A150241-250MG A150241-5G
76798-616EA 15.92 USD
76798-616 76798-618 76798-620 76798-622
Ethyl-3,6-dichloropyridazine-4-carboxylate 95%
Ethyl-3,6-dichloropyridazine-4-carboxylate

Precaución: for Research Use Only

Formula: C₇H₆Cl₂N₂O₂
MW: 221.04 g/mol
MDL Number: MFCD08704423
CAS Number: 34127-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR