DY131 98%

Supplier: Ambeed
Warning

Synonyms: N'-(4-(Diethylamino)benzylidene)-4-hydroxybenzohydrazide

A150000-10MG A150000-1MG A150000-25MG A150000-50MG
76798-588EA 19.1 USD
76798-588 76798-590 76798-592 76798-594
DY131 98%
DY131

Precaución: for Research Use Only

Formula: C₁₈H₂₁N₃O₂
MW: 311.38 g/mol
MDL Number: MFCD00715585
CAS Number: 95167-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR